Assay Details
Target Gene Details
Entrez Gene ID: | 63899 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 3 |
Gene Aliases: |
MST077, MSTP077 |
Location: |
Chr.3:94063011-94126786 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN3 | NM_022072.3 | NP_071355.1 | ||
AF169972.1 | AAQ13600.1 | |||
AK026262.1 | BAB15417.1 | |||
BC020602.1 | AAH20602.1 | |||
BC057238.1 | AAH57238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss | STX19 LOC107986037 ARL13B NSUN3 DHFR2 PROS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |