Assay Details
Target Gene Details
Entrez Gene ID: | 64084 |
Gene Name: | calsyntenin 2 |
Gene Aliases: |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
Location: |
Chr.3:139935185-140577397 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLSTN2 | NM_022131.2 | NP_071414.2 | ||
AJ278018.1 | CAC14924.1 | |||
AK096432.1 | ||||
AK315308.1 | ||||
AY753303.1 | AAV30553.1 | |||
BC103496.1 | AAI03497.1 | |||
BC103506.1 | AAI03507.1 | |||
BC104485.1 | AAI04486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829738 | Chr.3:139951994 - 140133103 on Build GRCh38 | Gain | CLSTN2 |
nsv998423 | Chr.3:139807604 - 140228494 on Build GRCh38 | Gain | CLSTN2 |
nsv591859 | Chr.3:139943096 - 139985332 on Build GRCh38 | Loss | CLSTN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |