Assay Details
Target Gene Details
Entrez Gene ID: | 27094 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 3 |
Gene Aliases: |
BKBETA3, HBETA3, K(VCA)BETA-3, KCNMB2, KCNMBL, SLO-BETA-3, SLOBETA3 |
Location: |
Chr.3:179239749-179267050 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMB3 | NM_001163677.1 | NP_001157149.1 | ||
NM_171828.2 | NP_741979.1 | |||
AK304837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010137 | Chr.3:179245109 - 179332999 on Build GRCh38 | Gain | ZNF639 KCNMB3 |
esv3893827 | Chr.3:179207079 - 179304740 on Build GRCh38 | Gain | PIK3CA KCNMB3 |
dgv5000n100 | Chr.3:179260056 - 179295493 on Build GRCh38 | Gain | KCNMB3 |
nsv829806 | Chr.3:179234575 - 179436585 on Build GRCh38 | Gain | MFN1 GNB4 PIK3CA ZNF639 KCNMB3 |
dgv4999n100 | Chr.3:179227018 - 179528391 on Build GRCh38 | Gain | MFN1 GNB4 PIK3CA ZNF639 KCNMB3 |
nsv592650 | Chr.3:179250846 - 179534475 on Build GRCh38 | Gain | MFN1 GNB4 ZNF639 KCNMB3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |