Assay Details
Target Gene Details
Entrez Gene ID: | 100529241 |
Gene Name: | HSPE1-MOB4 readthrough |
Gene Aliases: |
HSPE1-PHOCN |
Location: |
Chr.2:197499997-197553699 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPE1-MOB4 | NM_001202485.1 | NP_001189414.1 | ||
BJ992310.1 | ||||
CR990871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014881 | Chr.2:197432432 - 197668882 on Build GRCh38 | Gain | RFTN2 COQ10B SF3B1 HSPE1 HSPD1 MOB4 HSPE1-MOB4 SNORA105B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |