Assay Details
Target Gene Details
Entrez Gene ID: | 140739 |
Gene Name: | ubiquitin conjugating enzyme E2 F (putative) |
Gene Aliases: |
NCE2 |
Location: |
Chr.2:237966945-238042782 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2F | NM_001278305.1 | NP_001265234.1 | ||
NM_001278306.1 | NP_001265235.1 | |||
NM_001278307.1 | NP_001265236.1 | |||
NM_001278308.1 | NP_001265237.1 | |||
NM_080678.2 | NP_542409.1 | |||
NR_103498.1 | ||||
NR_103499.1 | ||||
NR_103500.1 | ||||
AF310723.1 | AAL26792.1 | |||
AK290063.1 | ||||
AK293334.1 | ||||
AK294107.1 | ||||
AK297502.1 | ||||
AK303094.1 | ||||
BC010549.1 | AAH10549.1 | |||
BM920749.1 | ||||
BQ959565.1 | ||||
BX421077.2 | ||||
DA140546.1 | ||||
DB118122.1 |
Target Gene Details
Entrez Gene ID: | 100533179 |
Gene Name: | UBE2F-SCLY readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:237966945-238099413 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2F-SCLY | NR_037904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | ASB18 LOC151174 UBE2F-SCLY MLPH RAB17 LINC01107 UBE2F PRLH COPS8 LOC105373958 LRRFIP1 ILKAP MIR6811 ACKR3 GBX2 SCLY RAMP1 LOC105373944 ESPNL RBM44 PER2 COL6A3 AGAP1 LOC101927958 ASB1 TRAF3IP1 LOC93463 KLHL30 ERFE IQCA1 HES6 LOC105373991 LOC643387 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |