Assay Details
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
METTL8 | NM_001321154.1 | 10 | 5553 | NP_001308083.1 |
NM_001321155.1 | 11 | 5677 | NP_001308084.1 | |
NM_001321156.1 | 11 | 5747 | NP_001308085.1 | |
NM_001321157.1 | 11 | 5580 | NP_001308086.1 | |
NM_001321158.1 | 9 | 5301 | NP_001308087.1 | |
NM_001321159.1 | 9 | 5398 | NP_001308088.1 | |
NM_001321160.1 | 10 | 5570 | NP_001308089.1 | |
NM_001321161.1 | 8 | 5194 | NP_001308090.1 | |
NM_001321162.1 | 9 | 5415 | NP_001308091.1 | |
NM_024770.4 | 10 | 5456 | NP_079046.2 | |
NR_135568.1 | 7 | 4798 | ||
XM_011511875.2 | 10 | 5368 | XP_011510177.1 | |
XM_017004984.1 | 11 | 5596 | XP_016860473.1 | |
XM_017004985.1 | 11 | 5694 | XP_016860474.1 | |
XM_017004986.1 | 10 | 5472 | XP_016860475.1 | |
XM_017004987.1 | 9 | 5334 | XP_016860476.1 | |
XM_017004988.1 | 9 | 5318 | XP_016860477.1 | |
XM_017004989.1 | 10 | 5487 | XP_016860478.1 | |
AK023396.1 | 1 | 932 | ||
AK055616.1 | 1 | 1726 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |