Assay Details
Target Gene Details
Entrez Gene ID: | 3694 |
Gene Name: | integrin subunit beta 6 |
Gene Aliases: |
AI1H |
Location: |
Chr.2:160099666-160200313 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB6 | NM_000888.4 | NP_000879.2 | ||
NM_001282353.1 | NP_001269282.1 | |||
NM_001282354.1 | NP_001269283.1 | |||
NM_001282355.1 | NP_001269284.1 | |||
NM_001282388.1 | NP_001269317.1 | |||
NM_001282389.1 | NP_001269318.1 | |||
NM_001282390.1 | NP_001269319.1 | |||
AK290300.1 | ||||
AK304203.1 | ||||
AK313944.1 | ||||
AL831998.1 | ||||
BC121178.2 | ||||
CD013944.1 | ||||
CD013945.1 | ||||
CD013946.1 | ||||
CD013947.1 | ||||
M35198.3 | AAA36122.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain | TANC1 MARCH7 PLA2R1 BAZ2B MIR6888 LOC643072 WDSUB1 LY75-CD302 LY75 LOC101929461 ITGB6 CD302 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |