Assay Details
Target Gene Details
Entrez Gene ID: | 57007 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
Location: |
Chr.2:236537131-236582358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACKR3 | XM_005246098.3 | XP_005246155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005677 | Chr.2:236369512 - 236811991 on Build GRCh38 | Gain | LOC105373991 IQCA1 ACKR3 |
nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain | LOC105373991 IQCA1 ASB18 ACKR3 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | RAB17 LOC643387 HES6 LOC105373958 LOC93463 PER2 MLPH LRRFIP1 SCLY RAMP1 GBX2 ASB1 ERFE LINC01107 ASB18 IQCA1 COPS8 AGAP1 LOC105373944 ESPNL ILKAP KLHL30 RBM44 ACKR3 LOC101927958 LOC105373991 MIR6811 PRLH UBE2F TRAF3IP1 UBE2F-SCLY LOC151174 COL6A3 |
nsv834576 | Chr.2:236466567 - 236654210 on Build GRCh38 | Gain | LOC105373991 IQCA1 ACKR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |