Assay Details
Target Gene Details
Entrez Gene ID: | 1593 |
Gene Name: | cytochrome P450 family 27 subfamily A member 1 |
Gene Aliases: |
CP27, CTX, CYP27 |
Location: |
Chr.2:218781006-218815293 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP27A1 | NM_000784.3 | NP_000775.1 | ||
XM_017003488.1 | XP_016858977.1 | |||
AK226039.1 | ||||
AK290418.1 | ||||
AK310782.1 | ||||
AY178622.1 | AAO21126.1 | |||
BC040430.1 | AAH40430.1 | |||
BC051851.1 | AAH51851.1 | |||
M62401.1 | AAA52142.1 | |||
X59812.1 | CAA42481.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |