Assay Details
Target Gene Details
Entrez Gene ID: | 91543 |
Gene Name: | radical S-adenosyl methionine domain containing 2 |
Gene Aliases: |
2510004L01Rik, cig33, cig5, vig1 |
Location: |
Chr.2:6865733-6898239 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RSAD2 | NM_080657.4 | NP_542388.2 | ||
XM_011510414.2 | XP_011508716.1 | |||
XM_011510415.2 | XP_011508717.1 | |||
AF442151.1 | AAL50053.1 | |||
AK300085.1 | ||||
AK312670.1 | ||||
BC017969.1 | AAH17969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss | RNF144A-AS1 RSAD2 RNF144A CMPK2 |
nsv2596 | Chr.2:6874902 - 6883625 on Build GRCh38 | Insertion | RSAD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |