Assay Details
Target Gene Details
Entrez Gene ID: | 101926982 |
Gene Name: | protein GVQW2-like |
Gene Aliases: |
- |
Location: |
Chr.2:9649308-9665708 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101926982 | XM_005246205.1 | XP_005246262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011802 | Chr.2:9626042 - 9668155 on Build GRCh38 | Gain | LOC105373418 LOC101926982 YWHAQ |
nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain | CPSF3 IAH1 LOC105373418 ITGB1BP1 LOC101926982 ASAP2 YWHAQ ADAM17 |
esv3589779 | Chr.2:9660410 - 9670694 on Build GRCh38 | Loss | LOC101926982 |
esv3589778 | Chr.2:9649326 - 9672251 on Build GRCh38 | Loss | LOC101926982 |
nsv527083 | Chr.2:9657865 - 9670519 on Build GRCh38 | Loss | LOC101926982 |