Assay Details
Target Gene Details
Entrez Gene ID: | 54842 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
MMR2, hMMR2 |
Location: |
Chr.2:190408355-190502315 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFSD6 | NM_017694.3 | NP_060164.3 | ||
XM_005246654.2 | XP_005246711.1 | |||
XM_005246656.3 | XP_005246713.1 | |||
XM_011511368.1 | XP_011509670.1 | |||
XM_017004392.1 | XP_016859881.1 | |||
AK127176.1 | BAC86869.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)