Assay Details
Target Gene Details
Entrez Gene ID: | 2825 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.2:206175316-206218047 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR1 | NM_001098199.1 | NP_001091669.1 | ||
NM_001261452.1 | NP_001248381.1 | |||
NM_001261453.1 | NP_001248382.1 | |||
NM_001261454.1 | NP_001248383.1 | |||
NM_001261455.1 | NP_001248384.1 | |||
NM_005279.3 | NP_005270.2 | |||
XM_005246471.2 | XP_005246528.1 | |||
XM_017003832.1 | XP_016859321.1 | |||
AK075130.1 | ||||
AK290976.1 | ||||
AK312917.1 | ||||
AL550780.3 | ||||
BC067833.1 | AAH67833.1 | |||
BP279126.1 | ||||
CB269800.1 | ||||
DA545892.1 | ||||
DA692381.1 | ||||
DA733626.1 | ||||
DA988428.1 |
Target Gene Details
Entrez Gene ID: | 101669764 |
Gene Name: | GPR1 antisense RNA |
Gene Aliases: |
GPR1-AS1, GPR1AS |
Location: |
Chr.2:206203376-206266243 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR1-AS | NR_104359.1 | |||
AB774455.1 | ||||
BE734382.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)