Assay Details
Target Gene Details
Entrez Gene ID: | 9378 |
Gene Name: | neurexin 1 |
Gene Aliases: |
Hs.22998, PTHSL2, SCZD17 |
Location: |
Chr.2:49918503-51032536 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)