Assay Details
Target Gene Details
Entrez Gene ID: | 9330 |
Gene Name: | general transcription factor IIIC subunit 3 |
Gene Aliases: |
TFIIIC102, TFIIICgamma, TFiiiC2-102 |
Location: |
Chr.2:196763032-196799768 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3C3 | NM_012086.4 | NP_036218.1 | ||
XM_005246965.4 | XP_005247022.1 | |||
AF085850.1 | ||||
AF133123.1 | AAD41475.1 | |||
AK025560.1 | ||||
AK027050.1 | BAB15638.1 | |||
AK314718.1 | ||||
AL137453.1 | CAB70745.1 | |||
BC015995.1 | AAH15995.1 | |||
BC043347.1 | AAH43347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575323 | Chr.2:196674745 - 197095661 on Build GRCh38 | Gain | LOC100130452 GTF3C3 ANKRD44 C2orf66 PGAP1 CCDC150 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |