Assay Details
Target Gene Details
Entrez Gene ID: | 10505 |
Gene Name: | ssemaphorin 4F |
Gene Aliases: |
M-SEMA, PRO2353, S4F, SEMAM, SEMAW, m-Sema-M |
Location: |
Chr.2:74654228-74699013 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA4F | NM_001271661.1 | NP_001258590.1 | ||
NM_001271662.1 | NP_001258591.1 | |||
NM_004263.4 | NP_004254.2 | |||
NR_073398.1 | ||||
NR_073399.1 | ||||
XM_011532475.2 | XP_011530777.1 | |||
XM_011532476.2 | XP_011530778.1 | |||
XM_011532477.2 | XP_011530779.1 | |||
XM_017003172.1 | XP_016858661.1 | |||
XM_017003173.1 | XP_016858662.1 | |||
XM_017003174.1 | XP_016858663.1 | |||
AB021292.1 | ||||
AB022317.1 | BAA75631.1 | |||
AK075384.1 | BAC11584.1 | |||
AK095587.1 | ||||
AK291023.1 | ||||
AK304358.1 | ||||
AK308166.1 | ||||
AK312728.1 | ||||
AL136552.1 | CAB66487.1 | |||
BC018361.1 | AAH18361.1 | |||
BC038411.1 | AAH38411.1 | |||
KU178531.1 | ||||
KU178532.1 | ||||
KU178533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003960 | Chr.2:74630228 - 74769504 on Build GRCh38 | Gain | M1AP SEMA4F |
nsv1001495 | Chr.2:74598036 - 75093218 on Build GRCh38 | Gain | LOC105374809 LINC01291 LINC01293 MIR5000 TACR1 M1AP POLE4 SEMA4F HK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |