Assay Details
Target Gene Details
Entrez Gene ID: | 55233 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
Location: |
Chr.2:74152598-74178967 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOB1A | NM_001317110.1 | NP_001304039.1 | ||
NM_001317111.1 | NP_001304040.1 | |||
NM_001317112.1 | NP_001304041.1 | |||
NM_018221.4 | NP_060691.2 | |||
XM_011532948.2 | XP_011531250.1 | |||
AB016839.1 | BAB19058.1 | |||
AJ577474.1 | CAE12093.1 | |||
AK001650.1 | BAA91810.1 | |||
AK023321.1 | BAB14525.1 | |||
BC003398.1 | AAH03398.1 | |||
BI825399.1 | ||||
BU172616.1 | ||||
CN426611.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)