Assay Details
Target Gene Details
Entrez Gene ID: | 65061 |
Gene Name: | cyclin dependent kinase 15 |
Gene Aliases: |
ALS2CR7, PFTAIRE2, PFTK2 |
Location: |
Chr.2:201790324-201895550 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDK15 | XM_005246782.4 | XP_005246839.1 | ||
AK292434.1 | ||||
AK310050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834511 | Chr.2:201777340 - 201964464 on Build GRCh38 | Loss | ALS2 CDK15 |
nsv524548 | Chr.2:201696848 - 201817506 on Build GRCh38 | Gain | MPP4 ALS2 CDK15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |