Assay Details
Target Gene Details
Entrez Gene ID: | 1593 |
Gene Name: | cytochrome P450 family 27 subfamily A member 1 |
Gene Aliases: |
CP27, CTX, CYP27 |
Location: |
Chr.2:218781006-218815293 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP27A1 | NM_000784.3 | NP_000775.1 | ||
XM_017003488.1 | XP_016858977.1 | |||
AK226039.1 | ||||
AK290418.1 | ||||
AK310782.1 | ||||
AY178622.1 | AAO21126.1 | |||
BC040430.1 | AAH40430.1 | |||
BC051851.1 | AAH51851.1 | |||
M62401.1 | AAA52142.1 | |||
X59812.1 | CAA42481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518750 | Chr.2:218751245 - 218804652 on Build GRCh38 | Loss | TTLL4 CYP27A1 |
esv3594349 | Chr.2:218756892 - 218789452 on Build GRCh38 | Loss | TTLL4 CYP27A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |