Assay Details
Target Gene Details
Entrez Gene ID: | 8828 |
Gene Name: | neuropilin 2 |
Gene Aliases: |
NP2, NPN2, PRO2714, VEGF165R2 |
Location: |
Chr.2:205682450-205798133 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834514 | Chr.2:205575978 - 205738443 on Build GRCh38 | Gain | PARD3B NRP2 |
nsv3123 | Chr.2:205681544 - 205714569 on Build GRCh38 | Insertion | NRP2 |
esv25775 | Chr.2:205407368 - 205806595 on Build GRCh38 | Gain | PARD3B NRP2 |
nsv460021 | Chr.2:205627674 - 205747861 on Build GRCh38 | Gain | NRP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |