Assay Details
Target Gene Details
Entrez Gene ID: | 80034 |
Gene Name: | cysteine and serine rich nuclear protein 3 |
Gene Aliases: |
FAM130A2, PPP1R73, TAIP-2, TAIP2 |
Location: |
Chr.2:165469647-165689407 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSRNP3 | NM_001172173.1 | NP_001165644.1 | ||
AK126129.1 | BAC86454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1015115 | Chr.2:165476091 - 165528603 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)