Assay Details
Target Gene Details
Entrez Gene ID: | 10267 |
Gene Name: | receptor activity modifying protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:237858714-237912117 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAMP1 | NM_001308353.1 | NP_001295282.1 | ||
NM_005855.3 | NP_005846.1 | |||
XM_017003152.1 | XP_016858641.1 | |||
XM_017003153.1 | XP_016858642.1 | |||
XM_017003155.1 | XP_016858644.1 | |||
XM_017003156.1 | XP_016858645.1 | |||
AI017108.1 | ||||
AJ001014.1 | CAA04472.1 | |||
AY265457.1 | AAP23298.1 | |||
BC000548.2 | AAH00548.1 | |||
BI548986.1 | ||||
CR542032.1 | CAG46829.1 | |||
CR542044.1 | CAG46841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584754 | Chr.2:237865062 - 237894836 on Build GRCh38 | Loss | RAMP1 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | LOC105373944 MIR6811 ASB1 KLHL30 PER2 AGAP1 IQCA1 LOC101927958 ESPNL ERFE LOC643387 ACKR3 LRRFIP1 PRLH LINC01107 LOC151174 ILKAP GBX2 RAB17 COPS8 UBE2F ASB18 COL6A3 RBM44 LOC105373991 MLPH HES6 RAMP1 SCLY LOC105373958 UBE2F-SCLY LOC93463 TRAF3IP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |