Assay Details
Target Gene Details
Entrez Gene ID: | 130752 |
Gene Name: | malate dehydrogenase 1B |
Gene Aliases: |
RP11-95H11 |
Location: |
Chr.2:206737763-206765549 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MDH1B | NM_001039845.2 | NP_001034934.1 | ||
NM_001282940.1 | NP_001269869.1 | |||
NR_104261.1 | ||||
XM_005246315.4 | XP_005246372.1 | |||
XM_005246316.4 | XP_005246373.1 | |||
XM_011510628.2 | XP_011508930.1 | |||
XM_017003373.1 | XP_016858862.1 | |||
XM_017003374.1 | XP_016858863.1 | |||
AK058070.1 | ||||
AK292378.1 | ||||
AK292386.1 | ||||
AK302250.1 | ||||
AK308257.1 | ||||
AK309144.1 | ||||
BC033509.1 | AAH33509.1 | |||
BC088356.1 | AAH88356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011347 | Chr.2:206384007 - 207085773 on Build GRCh38 | Gain | MDH1B ADAM23 MIR3130-2 DYTN FASTKD2 KLF7 LOC200726 MIR3130-1 CPO |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |