Assay Details
Target Gene Details
Entrez Gene ID: | 64965 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
MRP-S9, RPMS9, S9mt |
Location: |
Chr.2:105038025-105099960 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPS9 | NM_182640.2 | NP_872578.1 | ||
AK001451.1 | ||||
AK130078.1 | ||||
AV729381.1 | ||||
BC047784.1 | AAH47784.1 | |||
BC057240.1 | AAH57240.1 | |||
DT217261.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |