Assay Details
Target Gene Details
Entrez Gene ID: | 57574 |
Gene Name: | membrane associated ring-CH-type finger 4 |
Gene Aliases: |
MARCH-IV, RNF174 |
Location: |
Chr.2:216257862-216372027 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MARCH4 | NM_020814.2 | NP_065865.1 | ||
AB037820.1 | ||||
BC048793.1 | AAH48793.1 | |||
BC098448.1 | AAH98448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834537 | Chr.2:216266174 - 216448640 on Build GRCh38 | Loss | MARCH4 SMARCAL1 |
nsv1005697 | Chr.2:216231371 - 216429101 on Build GRCh38 | Loss | MARCH4 SMARCAL1 |
nsv1011506 | Chr.2:215969945 - 216310768 on Build GRCh38 | Gain | TMEM169 MARCH4 PECR PKI55 XRCC5 MREG |
nsv1003877 | Chr.2:216266863 - 216327849 on Build GRCh38 | Gain | MARCH4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |