Assay Details
Target Gene Details
Entrez Gene ID: | 842 |
Gene Name: | caspase 9 |
Gene Aliases: |
APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56 |
Location: |
Chr.1:15491401-15524912 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASP9 | NM_001229.4 | NP_001220.2 | ||
NM_001278054.1 | NP_001264983.1 | |||
NM_032996.3 | NP_127463.2 | |||
NR_102732.1 | ||||
NR_102733.1 | ||||
XM_005246014.2 | XP_005246071.1 | |||
XM_011542273.2 | XP_011540575.1 | |||
AB015653.1 | BAA78780.1 | |||
AB020979.1 | BAA87905.1 | |||
AB209147.1 | BAD92384.1 | |||
AF093130.1 | AAD12248.1 | |||
AF110376.1 | AAD13615.1 | |||
AJ781267.1 | CAH03131.1 | |||
AK292080.1 | ||||
AK292111.1 | ||||
AK303743.1 | ||||
AY732490.2 | AAV33129.1 | |||
BC002452.2 | AAH02452.1 | |||
BC006463.1 | AAH06463.1 | |||
BT006911.1 | AAP35557.1 | |||
DA078665.1 | ||||
HY027003.1 | ||||
U56390.1 | AAC50640.1 | |||
U60521.1 | AAC50776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss | CELA2A DNAJC16 FHAD1 CELA2B CTRC CASP9 EFHD2 LOC101927441 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |