Assay Details
Target Gene Details
Entrez Gene ID: | 56267 |
Gene Name: | kynurenine aminotransferase 3 |
Gene Aliases: |
CCBL2, KAT3, KATIII |
Location: |
Chr.1:88935773-88992960 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KYAT3 | NM_001008661.2 | NP_001008661.1 | ||
NM_001008662.2 | NP_001008662.1 | |||
AF091090.1 | AAC72959.1 | |||
AK057176.1 | ||||
AK301326.1 | ||||
AK315917.1 | ||||
AL833593.1 | ||||
AY028624.1 | AAK26163.1 | |||
BC000819.1 | AAH00819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546784 | Chr.1:88885058 - 88971430 on Build GRCh38 | Loss | GTF2B KYAT3 |
dgv207n100 | Chr.1:88932165 - 88995971 on Build GRCh38 | Loss | KYAT3 RBMXL1 |
nsv830525 | Chr.1:88776863 - 88975647 on Build GRCh38 | Loss | GTF2B PKN2 KYAT3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |