Assay Details
Target Gene Details
Entrez Gene ID: | 64744 |
Gene Name: | small ArfGAP2 |
Gene Aliases: |
SMAP1L |
Location: |
Chr.1:40373706-40423326 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMAP2 | NM_001198978.1 | NP_001185907.1 | ||
NM_022733.2 | NP_073570.1 | |||
XM_011541960.1 | XP_011540262.1 | |||
XM_017002093.1 | XP_016857582.1 | |||
AK303975.1 | ||||
AK313105.1 | ||||
AL133206.1 | CAB61580.1 | |||
AL137764.1 | CAB70912.1 | |||
BC021133.1 | AAH21133.1 | |||
DC411665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585803 | Chr.1:40375034 - 40406101 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)