Assay Details
Target Gene Details
Entrez Gene ID: | 79033 |
Gene Name: | ERI1 exoribonuclease family member 3 |
Gene Aliases: |
PINT1, PRNPIP |
Location: |
Chr.1:44221070-44355298 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERI3 | NM_001301698.1 | NP_001288627.1 | ||
NM_001301699.1 | NP_001288628.1 | |||
NM_001301700.1 | NP_001288629.1 | |||
NM_001301701.1 | NP_001288630.1 | |||
NM_024066.2 | NP_076971.1 | |||
XM_005271186.2 | XP_005271243.1 | |||
XM_006710891.3 | XP_006710954.1 | |||
XM_017002301.1 | XP_016857790.1 | |||
XM_017002302.1 | XP_016857791.1 | |||
XM_017002303.1 | XP_016857792.1 | |||
XM_017002304.1 | XP_016857793.1 | |||
XM_017002305.1 | XP_016857794.1 | |||
AF007157.1 | AAC19158.1 | |||
AK026596.1 | ||||
AK290567.1 | ||||
AK293833.1 | ||||
AK297708.1 | ||||
BC001072.2 | AAH01072.1 | |||
BC004456.1 | AAH04456.1 | |||
BI765887.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |