Assay Details
Target Gene Details
Entrez Gene ID: | 374946 |
Gene Name: | dorsal inhibitory axon guidance protein |
Gene Aliases: |
AGPA3119, C1orf187, UNQ3119, neucrin |
Location: |
Chr.1:11686635-11721766 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DRAXIN | NM_198545.3 | NP_940947.3 | ||
XM_011541405.2 | XP_011539707.1 | |||
XM_017001214.1 | XP_016856703.1 | |||
XM_017001215.1 | XP_016856704.1 | |||
AK075558.1 | BAC11697.1 | |||
AY358750.1 | AAQ89110.1 | |||
BC111064.1 | AAI11065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832758 | Chr.1:11637026 - 11777087 on Build GRCh38 | Loss | AGTRAP MAD2L2 FBXO6 FBXO44 LOC102724659 C1orf167 FBXO2 DRAXIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |