Assay Details
Target Gene Details
Entrez Gene ID: | 2060 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 |
Gene Aliases: |
AF-1P, AF1P, MLLT5 |
Location: |
Chr.1:51354263-51519354 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS15 | NM_001159969.1 | NP_001153441.1 | ||
NM_001981.2 | NP_001972.1 | |||
XM_005270618.2 | XP_005270675.1 | |||
XM_017000615.1 | XP_016856104.1 | |||
XM_017000616.1 | XP_016856105.1 | |||
XM_017000617.1 | XP_016856106.1 | |||
XM_017000618.1 | XP_016856107.1 | |||
XM_017000619.1 | XP_016856108.1 | |||
XM_017000620.1 | XP_016856109.1 | |||
AK090936.1 | ||||
AK294297.1 | ||||
AK308128.1 | ||||
AK313396.1 | ||||
BC054006.1 | AAH54006.1 | |||
BX647676.1 | ||||
DQ367924.1 | ||||
U07707.1 | AAA52101.1 | |||
Z29064.1 | CAA82305.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |