Assay Details
Target Gene Details
Entrez Gene ID: | 117145 |
Gene Name: | thioesterase superfamily member 4 |
Gene Aliases: |
CTMP |
Location: |
Chr.1:151870866-151909885 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THEM4 | NM_053055.4 | NP_444283.2 | ||
XM_011509142.2 | XP_011507444.1 | |||
AJ313515.1 | CAC86384.1 | |||
AK289494.1 | ||||
AK314852.1 | ||||
BC065277.1 | AAH65277.1 | |||
BE349541.1 | ||||
BM787449.1 | ||||
DC353229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv547852 | Chr.1:151870568 - 151982754 on Build GRCh38 | Loss | THEM4 |
nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain | RPTN S100A11 NBPF18P S100A10 TCHH LOC100131107 THEM4 TCHHL1 HRNR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |