Assay Details
Target Gene Details
Entrez Gene ID: | 2475 |
Gene Name: | mechanistic target of rapamycin |
Gene Aliases: |
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS |
Location: |
Chr.1:11106531-11262551 on Build GRCh38 |
Assay Gene Location: | Within Intron 55 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTOR | NM_004958.3 | NP_004949.1 | ||
XM_005263438.2 | XP_005263495.1 | |||
XM_017000900.1 | XP_016856389.1 | |||
XM_017000901.1 | XP_016856390.1 | |||
AB209995.1 | ||||
AK126762.1 | ||||
AK302863.1 | ||||
AK304273.1 | ||||
BC117166.1 | ||||
L34075.1 | AAA58486.1 | |||
U88966.1 | AAC39933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain | EXOSC10 TARDBP LOC105376736 MASP2 SRM MTOR-AS1 UBIAD1 ANGPTL7 MTOR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |