Assay Details
Target Gene Details
Entrez Gene ID: | 1650 |
Gene Name: | dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit |
Gene Aliases: |
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1 |
Location: |
Chr.1:20651767-20661544 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDOST | NM_005216.4 | NP_005207.2 | ||
AB062391.1 | BAB93478.1 | |||
AK296041.1 | ||||
AK297009.1 | ||||
AK315633.1 | ||||
BC002594.2 | AAH02594.1 | |||
BP233191.1 | ||||
CB988524.1 | ||||
D29643.1 | BAA06126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1003348 | Chr.1:20648970 - 20715062 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv834369 | Chr.1:20609770 - 20779837 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)