Assay Details
Target Gene Details
Entrez Gene ID: | 80896 |
Gene Name: | N-acetylneuraminate pyruvate lyase |
Gene Aliases: |
C112, C1orf13, NAL, NPL1 |
Location: |
Chr.1:182789449-182830384 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPL | NM_001200050.1 | NP_001186979.1 | ||
NM_001200051.1 | NP_001186980.1 | |||
NM_001200052.1 | NP_001186981.1 | |||
NM_001200056.1 | NP_001186985.1 | |||
NM_030769.2 | NP_110396.1 | |||
AF338436.1 | AAK25795.1 | |||
AK297017.1 | ||||
AK301068.1 | ||||
AK313225.1 | ||||
AL525867.3 | ||||
AY336748.1 | AAQ82432.1 | |||
BC058003.1 | AAH58003.1 | |||
BC125051.1 | ||||
CB961482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain | RGSL1 TEDDM1 NPL RNASEL LOC105371644 DHX9 LAMC1 LOC647070 ZNF648 RGS16 LINC00272 LINC01344 RGS8 SHCBP1L LOC284648 GLUL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |