Assay Details
Target Gene Details
Entrez Gene ID: | 54482 |
Gene Name: | tRNA methyltransferase 13 homolog |
Gene Aliases: |
CCDC76 |
Location: |
Chr.1:100133080-100150498 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRMT13 | NM_019083.2 | NP_061956.2 | ||
NR_135078.1 | ||||
XM_005270945.3 | XP_005271002.1 | |||
XM_005270946.2 | XP_005271003.1 | |||
XM_017001504.1 | XP_016856993.1 | |||
XM_017001505.1 | XP_016856994.1 | |||
AK001149.1 | BAA91520.1 | |||
AK002081.1 | BAA92074.1 | |||
AK289404.1 | ||||
AK294738.1 | ||||
AK295828.1 | ||||
AK298941.1 | ||||
AL699548.1 | ||||
BC075811.1 | AAH75811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2121 | Chr.1:100115063 - 100149647 on Build GRCh38 | Insertion | LRRC39 TRMT13 SASS6 |
nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain | LRRC39 TRMT13 DBT MFSD14A SASS6 |
esv275107 | Chr.1:100136209 - 100139757 on Build GRCh38 | Loss | TRMT13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |