Assay Details
Target Gene Details
Entrez Gene ID: | 51603 |
Gene Name: | methyltransferase like 13 |
Gene Aliases: |
5630401D24Rik, CGI-01, KIAA0859, feat |
Location: |
Chr.1:171781504-171823962 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
METTL13 | NM_001007239.1 | NP_001007240.1 | ||
NM_014955.2 | NP_055770.1 | |||
NM_015935.4 | NP_057019.3 | |||
XM_017001455.1 | XP_016856944.1 | |||
XM_017001456.1 | XP_016856945.1 | |||
XM_017001457.1 | XP_016856946.1 | |||
XM_017001458.1 | XP_016856947.1 | |||
XM_017001459.1 | XP_016856948.1 | |||
AB020666.2 | ||||
AB242174.1 | ||||
AB242175.1 | ||||
AF132936.1 | AAD27711.1 | |||
AK001172.1 | ||||
AK027552.1 | ||||
AK027621.1 | BAB55240.1 | |||
AK027886.1 | ||||
AK074552.1 | BAC11055.1 | |||
AK129561.1 | ||||
AK223493.1 | BAD97213.1 | |||
AK291740.1 | ||||
AK304471.1 | ||||
AL049669.1 | CAB41243.1 | |||
BC006814.1 | AAH06814.1 | |||
BC029083.1 | AAH29083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518222 | Chr.1:171783899 - 171794409 on Build GRCh38 | Loss | METTL13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |