Assay Details
Target Gene Details
Entrez Gene ID: | 1727 |
Gene Name: | cytochrome b5 reductase 3 |
Gene Aliases: |
B5R, DIA1 |
Location: |
Chr.22:42617840-42649399 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYB5R3 | NM_000398.6 | NP_000389.1 | ||
NM_001129819.2 | NP_001123291.1 | |||
NM_001171660.1 | NP_001165131.1 | |||
NM_001171661.1 | NP_001165132.1 | |||
NM_007326.4 | NP_015565.1 | |||
AF361370.1 | AAL87744.1 | |||
AJ010116.1 | CAA09006.1 | |||
AJ010117.1 | CAA09007.1 | |||
AJ010118.1 | CAA09008.1 | |||
AJ310899.1 | CAC84523.1 | |||
AJ310900.1 | CAC84524.1 | |||
AK124528.1 | BAC85875.1 | |||
AK302204.1 | ||||
BC004821.1 | AAH04821.1 | |||
BM564146.1 | ||||
BT009821.1 | AAP88823.1 | |||
CR456435.1 | CAG30321.1 | |||
M16462.1 | AAA52307.1 | |||
Y09501.1 | CAA70696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524028 | Chr.22:42592111 - 42631818 on Build GRCh38 | Loss | POLDIP3 RNU12 CYB5R3 |
esv2758839 | Chr.22:42371195 - 42687300 on Build GRCh38 | Gain+Loss | POLDIP3 RRP7A RRP7BP SERHL2 ATP5L2 NFAM1 RNU12 CYB5R3 SERHL |
nsv428391 | Chr.22:42371295 - 42698202 on Build GRCh38 | Gain | POLDIP3 RRP7A RRP7BP SERHL2 ATP5L2 NFAM1 RNU12 A4GALT CYB5R3 SERHL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |