Assay Details
Target Gene Details
Entrez Gene ID: | 79016 |
Gene Name: | DET1 and DDB1 associated 1 |
Gene Aliases: |
C19orf58, PCIA1 |
Location: |
Chr.19:17309528-17323301 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDA1 | NM_024050.5 | NP_076955.1 | ||
AY563006.2 | AAS66629.1 | |||
BC000615.2 | AAH00615.1 | |||
DQ090952.1 | AAZ41375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss | GTPBP3 MRPL34 ANKLE1 OCEL1 ABHD8 ANO8 BABAM1 NR2F6 USHBP1 MYO9B DDA1 USE1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |