Assay Details
Target Gene Details
Entrez Gene ID: | 220164 |
Gene Name: | docking protein 6 |
Gene Aliases: |
DOK5L, HsT3226 |
Location: |
Chr.18:69401027-69849087 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOK6 | NM_152721.5 | NP_689934.2 | ||
XM_011525875.2 | XP_011524177.1 | |||
AY599248.1 | AAT09770.1 | |||
BC096744.1 | AAH96744.1 | |||
DB480182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3642965 | Chr.18:69412395 - 69428741 on Build GRCh38 | Loss |
![]() |
esv32736 | Chr.18:69135149 - 71678611 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3642964 | Chr.18:69398892 - 69788959 on Build GRCh38 | Loss |
![]() ![]() |
nsv523105 | Chr.18:69419384 - 69477833 on Build GRCh38 | Gain |
![]() ![]() |
esv2717289 | Chr.18:69412388 - 69428685 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)