Assay Details
Target Gene Details
Entrez Gene ID: | 100506677 |
Gene Name: | uncharacterized LOC100506677 |
Gene Aliases: |
AA02, AA04, AA05 |
Location: |
Chr.17:33529787-33533760 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AA06 | NR_037584.2 | |||
AF381171.1 | ||||
AF381172.1 | ||||
AF381173.1 | ||||
AF381174.1 |
Target Gene Details
Entrez Gene ID: | 40 |
Gene Name: | acid sensing ion channel subunit 2 |
Gene Aliases: |
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1 |
Location: |
Chr.17:33013087-34156806 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASIC2 | NM_001094.4 | NP_001085.2 | ||
AK313043.1 | ||||
BC075042.2 | AAH75042.1 | |||
BC075043.2 | AAH75043.1 | |||
U50352.1 | AAC50432.1 | |||
U53212.1 | AAC50498.1 | |||
U57352.1 | AAB49182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521841 | Chr.17:33406743 - 33593135 on Build GRCh38 | Loss | AA06 LOC147004 ASIC2 |
nsv9539 | Chr.17:33358415 - 33598698 on Build GRCh38 | Loss | AA06 LOC147004 ASIC2 |
nsv833421 | Chr.17:33520714 - 33663203 on Build GRCh38 | Gain | AA06 LOC147004 ASIC2 |
nsv510706 | Chr.17:33437824 - 33596377 on Build GRCh38 | Deletion | AA06 LOC147004 ASIC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |