Assay Details
Target Gene Details
Entrez Gene ID: | 123096 |
Gene Name: | solute carrier family 25 member 29 |
Gene Aliases: |
C14orf69, CACL, ORNT3 |
Location: |
Chr.14:100277986-100306547 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635502 | Chr.14:100275457 - 100309132 on Build GRCh38 | Gain | SLC25A29 YY1 MIR345 MIR6764 |
nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss | SLC25A29 YY1 MIR345 SLC25A47 WARS MIR6764 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |