Assay Details
Target Gene Details
Entrez Gene ID: | 7347 |
Gene Name: | ubiquitin C-terminal hydrolase L3 |
Gene Aliases: |
UCH-L3 |
Location: |
Chr.13:75549480-75606020 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UCHL3 | NM_001270952.1 | NP_001257881.1 | ||
NM_006002.4 | NP_005993.1 | |||
XM_011535212.1 | XP_011533514.1 | |||
XM_011535213.2 | XP_011533515.1 | |||
XM_011535214.2 | XP_011533516.1 | |||
XM_017020725.1 | XP_016876214.1 | |||
XM_017020726.1 | XP_016876215.1 | |||
XM_017020727.1 | XP_016876216.1 | |||
AK313665.1 | ||||
BC018125.2 | AAH18125.1 | |||
BQ427804.1 | ||||
BT019359.1 | AAV38166.1 | |||
CR456855.1 | CAG33136.1 | |||
M30496.1 | AAA36791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss | UCHL3 LMO7-AS1 LMO7 |
nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain | LOC105370259 LINC00381 UCHL3 COMMD6 LMO7-AS1 LINC00347 KLF12 LMO7 LINC01078 CTAGE11P TBC1D4 |
esv2761803 | Chr.13:75369173 - 75587638 on Build GRCh38 | Gain | UCHL3 COMMD6 TBC1D4 |
nsv562347 | Chr.13:75558104 - 75579279 on Build GRCh38 | Loss | UCHL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |