Assay Details
Target Gene Details
Entrez Gene ID: | 4254 |
Gene Name: | KIT ligand |
Gene Aliases: |
DCUA, DFNA69, FPH2, FPHH, KL-1, Kitl, MGF, SCF, SF, SHEP7 |
Location: |
Chr.12:88492793-88580473 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KITLG | NM_000899.4 | NP_000890.1 | ||
NM_003994.5 | NP_003985.2 | |||
AF400436.1 | AAK92485.1 | |||
AK290319.1 | ||||
AK293002.1 | ||||
BC069733.1 | AAH69733.1 | |||
BC069783.1 | AAH69783.1 | |||
BC069797.1 | AAH69797.1 | |||
BC126166.1 | ||||
BC143899.1 | ||||
CR749222.1 | CAH18078.1 | |||
M59964.1 | AAA85450.1 | |||
S45995.1 | AAB23573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483111 | Chr.12:86306223 - 88606223 on Build GRCh38 | Loss | C12orf50 TMTC3 CEP290 LOC105369879 MKRN9P MGAT4C KITLG C12orf29 |
nsv559704 | Chr.12:88372790 - 88645023 on Build GRCh38 | Gain | KITLG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |