Assay Details
Target Gene Details
Entrez Gene ID: | 8453 |
Gene Name: | cullin 2 |
Gene Aliases: |
- |
Location: |
Chr.10:35008551-35126899 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CUL2 | NM_001198777.1 | NP_001185706.1 | ||
NM_001198778.1 | NP_001185707.1 | |||
NM_001198779.1 | NP_001185708.1 | |||
NM_001324375.1 | NP_001311304.1 | |||
NM_001324376.1 | NP_001311305.1 | |||
NM_003591.3 | NP_003582.2 | |||
XM_011519743.1 | XP_011518045.1 | |||
XM_011519744.1 | XP_011518046.1 | |||
XM_011519745.1 | XP_011518047.1 | |||
XM_011519747.1 | XP_011518049.1 | |||
AF126404.1 | AAD23581.1 | |||
AK095217.1 | ||||
AK294080.1 | ||||
AK300491.1 | ||||
AK310225.1 | ||||
AL832733.1 | ||||
BC009591.1 | AAH09591.1 | |||
BC110901.1 | AAI10902.1 | |||
U58088.1 | AAC50545.1 | |||
U83410.1 | AAC51190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv550401 | Chr.10:34931247 - 35060646 on Build GRCh38 | Gain | CUL2 |
nsv550391 | Chr.10:34383535 - 35056041 on Build GRCh38 | Gain | PARD3-AS1 CUL2 PARD3 |
nsv466870 | Chr.10:35011695 - 35049771 on Build GRCh38 | Loss | CUL2 |
dgv705n100 | Chr.10:34871114 - 35045560 on Build GRCh38 | Gain | CUL2 |
dgv202e212 | Chr.10:34880967 - 35026718 on Build GRCh38 | Gain | CUL2 |
nsv466869 | Chr.10:34966937 - 35052373 on Build GRCh38 | Gain | CUL2 |
dgv706n100 | Chr.10:34881487 - 35230186 on Build GRCh38 | Gain | CUL2 CREM MIR3611 |
nsv550398 | Chr.10:34885707 - 35271661 on Build GRCh38 | Gain | CCNY CUL2 CREM MIR3611 |
nsv1161937 | Chr.10:34971581 - 35049745 on Build GRCh38 | Duplication | CUL2 |
dgv703n100 | Chr.10:34729183 - 35050396 on Build GRCh38 | Gain | PARD3-AS1 CUL2 PARD3 |
nsv470931 | Chr.10:34695956 - 35060646 on Build GRCh38 | Gain | PARD3-AS1 CUL2 PARD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |