Assay Details
Target Gene Details
Entrez Gene ID: | 79269 |
Gene Name: | DDB1 and CUL4 associated factor 10 |
Gene Aliases: |
WDR32 |
Location: |
Chr.9:37800554-37867669 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF10 | NM_001286810.1 | NP_001273739.1 | ||
NM_024345.4 | NP_077321.3 | |||
XM_005251577.3 | XP_005251634.1 | |||
XM_005251578.3 | XP_005251635.1 | |||
XM_017015128.1 | XP_016870617.1 | |||
XM_017015129.1 | XP_016870618.1 | |||
AK074532.1 | BAC11043.1 | |||
AK309712.1 | ||||
AK315756.1 | ||||
AL533994.3 | ||||
BC108686.1 | AAI08687.1 | |||
BC139834.1 | ||||
CD299973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831552 | Chr.9:37693652 - 37859980 on Build GRCh38 | Gain | EXOSC3 TRMT10B LOC105376037 DCAF10 FRMPD1 |
nsv469814 | Chr.9:37777286 - 37950154 on Build GRCh38 | Gain | EXOSC3 TRMT10B LOC105376037 SLC25A51 DCAF10 SHB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |