Assay Details
Target Gene Details
Entrez Gene ID: | 23484 |
Gene Name: | leptin receptor overlapping transcript-like 1 |
Gene Aliases: |
HSPC112, Vps55, my047 |
Location: |
Chr.8:30095406-30137706 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LEPROTL1 | NM_001128208.1 | NP_001121680.1 |
Target Gene Details
Entrez Gene ID: | 619373 |
Gene Name: | membrane bound O-acyltransferase domain containing 4 |
Gene Aliases: |
FKSG89, GOAT, OACT4 |
Location: |
Chr.8:30131454-30148768 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT4 | NM_001100916.1 | NP_001094386.1 | ||
XM_017013726.1 | XP_016869215.1 | |||
EU518495.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831277 | Chr.8:29977428 - 30180297 on Build GRCh38 | Gain | MBOAT4 LEPROTL1 SARAF DCTN6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |