Assay Details
Target Gene Details
Entrez Gene ID: | 10434 |
Gene Name: | lysophospholipase I |
Gene Aliases: |
APT-1, APT1, LPL-I, LPL1, hAPT1 |
Location: |
Chr.8:54046367-54102017 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576573 | Chr.8:53946867 - 54052964 on Build GRCh38 | Gain | LOC105375839 LYPLA1 TCEA1 RGS20 |
esv2759617 | Chr.8:53920430 - 54104405 on Build GRCh38 | Gain | LOC105375839 LYPLA1 TCEA1 RGS20 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |