Assay Details
Target Gene Details
Entrez Gene ID: | 100996348 |
Gene Name: | CA3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:85441806-85464915 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA3-AS1 | NR_121630.1 | |||
NR_121631.1 | ||||
NR_121632.1 | ||||
NR_121633.1 | ||||
AW172333.1 | ||||
AW611902.1 | ||||
BM713112.1 | ||||
BM717037.1 | ||||
DA940436.1 | ||||
DB177224.1 | ||||
DB196812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831377 | Chr.8:85350878 - 85531912 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1028148 | Chr.8:85398341 - 85640901 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2761206 | Chr.8:85398353 - 85556639 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)