Assay Details
Target Gene Details
Entrez Gene ID: | 6863 |
Gene Name: | tachykinin precursor 1 |
Gene Aliases: |
Hs.2563, NK2, NKNA, NPK, TAC2 |
Location: |
Chr.7:97731959-97740472 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAC1 | NM_003182.2 | NP_003173.1 | ||
NM_013996.2 | NP_054702.1 | |||
NM_013997.2 | NP_054703.1 | |||
NM_013998.2 | NP_054704.1 | |||
AF050655.1 | AAC15701.1 | |||
AF050656.1 | AAC15702.1 | |||
AF050657.1 | AAC15703.1 | |||
AF050658.1 | AAC15704.1 | |||
AK222957.1 | BAD96677.1 | |||
BC018047.1 | AAH18047.1 | |||
CR407602.1 | CAG28529.1 | |||
CR541730.1 | CAG46531.1 | |||
DN992454.1 | ||||
M68906.1 | AAA60159.1 | |||
M68907.1 | AAA60160.1 | |||
U37529.1 | AAA79195.1 | |||
X54469.1 | CAA38351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv437559 | Chr.7:97732472 - 97754008 on Build GRCh38 | Loss | TAC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |